Lumitester smart, Kikkoman
Toimittaja: HYSERVE
1702671 1702672 1902653
710-2244EA 375 EUR
710-2244 710-2245 710-2243
Lumitester smart, Kikkoman
Valvontajärjestelmät Hygieniavalvonta
Luminometer for hygiene validation of surfaces in 10 seconds.
The lumitester Smart, in combination with the LuciPac A3 reagent tests, detects all organic residues and microorganisms on surfaces as well as in liquids and determines the degree of cleanliness at the point of sampling. The system thus enables hygiene monitoring of surfaces within seconds, directly following cleaning and disinfection measures.
The Lumitester Smart is a mobile device that allows it to be used even in places that are difficult to access, making it particularly user-friendly. Via Bluetooth, the device can beMore Product Information