Air sampler, Coriolis®
Toimittaja: Bertin Instruments
P001080-CORM0A
710-1973EA 9890 EUR
710-1973
Air sampler, Coriolis®
Näytteenottimet Ilmanäytekerääjät
Coriolis® μ is a portable biological air sampler for indoor or outdoor air bio-contamination monitoring. The equipment concentrates airborne particles in a liquid sample that can be analysed afterwards using microbiological techniques.
  • Concentrates airborne particles in a liquid sample
  • Compatible with culture and molecular biology standard methods (qPCR included)
  • Easy decontamination (hydrogen peroxide)
  • Autonomy on battery of 1 hour
  • Optional longer collection system up to 6 hours
Sample types include toxins, virus, bacteria, moulds, pollens and spores.More Product Information