Real-Time PCR kit for detection of Legionella spec.
Toimittaja: Q BIOANALYTIC
QB-RTI-16-1000 QB-RTI-16-250 QB-RTI-16-50
710-1757EA 0 EUR
710-1757 710-1759 710-1760
Real-Time PCR kit for detection of Legionella spec.
Pikatestit patogeenien ja allergeenien osoittamiseen PCR testit
Legionella spec. is tested usually with cultural methods. However, this may take up to 14 days to deliver results and some strains are living but are not culturable. Using this Real-Time PCR test you may have results already after a few hours. You can use this test in combination with an appropriate pre- enrichment or you can do a DNA purification directly from the filter without pre-enrichment. The kit is also suitable to confirm bacterial colonies on agar plates.
  • TaqMan probes
  • Contains internal amplification control in HEX channel
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Sertifikaatit: DIN EN ISO 20838 conform detection in water samples from pre-enrichment cultures or for confirmation of bacterial colonies.