VWR® Contact Plate Weight Applicator, Contact Weight
Toimittaja: VWR Collection
710-0961EA 454 EUR
710-0961
VWR® Contact Plate Weight Applicator, Contact Weight
Elatusaineet mikrobiologiaan Kontaktimaljat mikrobiologiaan
Used in combination with contact plates, the contact weight enables the standardisation of microbiological surface control between operators. It applies the same constant pressure for a pre-set time on a contact plate. This ensures results are directly comparable. The contact weight meets the ISO 18593 standard, and has a sterilisable stainless steel base and pull-out digital timer.
  • Ruostumattomasta teräksestä valmistetun jalustan voi steriloida kuumailmalla tai höyryllä. Mukana digitaalinen, ulos vedettävä ajastin
  • Soveltuu monenlaisilla kontaktimaljaille
  • Standardin ISO 18593 vaatimusten mukainen
  • Painaa vain 500 g
Timer included provided without the certificate of calibration; if a calibration is required it is necessary to buy the article 609-0184 as accessory that includes the calibration.