Count plates, Petrifilm™
Toimittaja: 3M Food Safety
6490 6491 6492 6493 6407 6410 6417 6434 6435 6450
710-0777EA 125 EUR
710-0777 710-0778 710-0779 710-0780 710-3408 710-0770 710-3409 710-3406 710-3407 710-0849
Count plates, Petrifilm™
Elatusaineet mikrobiologiaan Valmisteltu viljelyaine
3M™ Petrifilm™ is an extremely cost-effective, practical and reliable method for testing equipment, raw materials, food products and their manufacturing environment; it also enables more frequent monitoring of critical control points. Fast and easy-to-approve results provide you with the information required to make the right decisions. The end result is an increase in product quality, improved process control and an overall reduction in operating costs.
  • Fast and accurate results in 48 hours
  • Space-saving
  • 3M™ Petrifilm™ plates can also be read in four seconds using the 3M™ Petrifilm™ plate reader