Real-Time PCR kit for detection of pork meat in food samples
Toimittaja: Q BIOANALYTIC
QB-RT-13-10SMP QB-RT-13-50 QB-RT-13-250 QB-RT-13-1000
710-0698EA 0 EUR
710-0698 710-0704 710-0699 710-0697
Real-Time PCR kit for detection of pork meat in food samples
Pikatestit patogeenien ja allergeenien osoittamiseen PCR testit
Sus scrofa (pork) halal and kosher food does exclude pork meet. To make sure that pork meat is really absent in complex food it is advisable to test for the presence of it using our Real-Time test. Positive DNA and all required components are included in the kit.
  • TaqMan probes
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included