Small compressed head swab, CleanFoam®
Toimittaja: Texwipe
TX751B
710-0576EA 96 EUR
710-0576
Small compressed head swab, CleanFoam®
Vanupuikot Näytteenottopuikot puhdistamiseen
Excellent swab for cleaning very small areas, especially thin, grooved areas of parts and assemblies.
  • Very low particle generation
  • Small compressed CleanFoam head with pointed tip on a short rigid handle
  • Polyurethane foam head has been solvent washed to greatly reduce non volatile residue
Suitable for precision application of solvents and cleaning solutions.
Pakkaus: Cleanroom processed bag-within-a-bag.
Varoitus: *Tällä sivulla olevat tuotteet eivät kuulu EU -direktiivin 2019/904 soveltamisalaan