Vanupuikot, Constix®, pointed foam head
Toimittaja: CONTEC
SF-13ESD
710-0573EA 248 EUR
710-0573
Vanupuikot, Constix®, pointed foam head
Vanupuikot Näytteenottopuikot puhdistamiseen
This static-safe swab consists of a pointed-tip sealed foam head attached to a conductive polymer handle.
  • ESD handle is great for static-sensitive applications
  • Fine point enables access to very small areas and crevices
  • Polyurethane foam head provides good particle entrapment and removal
  • Contains no binders or adhesives
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery and removal.
The SF-13ESD is also compatible and sorbent with most common solvents. NotMore Product Information