J.T.Baker® BAKERBOND spe™ Quaternary Amine, SPE Columns
Toimittaja: Avantor
7091-01EA 357.54 EUR
7091-01 7091-03 7091-07
J.T.Baker® BAKERBOND spe™ Quaternary Amine, SPE Columns
Kromatografiakolonnit
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Ionic, acidic analytes.
These SPE columns can be used in strong anion exchange mode.