J.T.Baker® BAKERBOND spe™ SiOH, SPE Columns
Toimittaja: Avantor
7086-03EA 202 EUR
7086-03 7086-01 7086-06 7086-02 7086-23 7086-26 7337-08 7086-08 7337-07 7086-07
J.T.Baker® BAKERBOND spe™ SiOH, SPE Columns
Kromatografiakolonnit
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Polar analytes from non polar solvents like hydrocarbons and less polar esters and ethers.
These SPE columns can be used in adsorption mode.
Pakkaus: Certain columns are available in a foil-lined fibre drum called the Jumbo Pack. TheMore Product Information