Kloroformi, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor
Vaara

7071.2500EA 245.73 EUR
7071.2500 7071.1000
Kloroformi, BAKER ANALYZED® ACS, J.T.Baker®
Kloroformi
Kaava: CHCl₃
Kiehumispiste: 61,7 °C (1013 hPa)
Sulamispiste: –63 °C
Tiheys: 1,4832 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000826
CAS-numero: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Exceeds ACS Specifications
Assay (by GC) min. 99.8 %
Acetone and Aldehyde (as (CH3)2CO) max. 0.001 %
Acid and Chloride passes test
Color (APHA) max. 10
Lead (Pb) max. 0.05 ppm
Free Chlorine passes test
Preservative (C2H5OH) about 0.75%
Residue after Evaporation max. 5 ppm
Substances Darkened by H2SO4 passes test
Suitability for Use in Dithizone Test passes test
Ultraviolet Absorbance (1.00-cm path vs water):
at 245 nm max. 1.00
at 255 nm max. 0.25
at 260 nm max. 0.15
at 270 nm max. 0.05
at 290-400 nm max. 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR