Kaliumhydroksidi 45% vesiliuos, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Toimittaja: Avantor
Vaara

7063.1000EA 60.28 EUR
7063.1000 7063.9025
Kaliumhydroksidi 45% vesiliuos, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Kaliumhydroksidi
Kaava: KOH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003553
CAS-numero: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now

Testitulosten erittely

Assay min. 45.0 %
Ammonium Hydroxide Precipitate max. 0.005 %
Chloride (Cl) max. 0.005 %
Potassium Carbonate (K2CO3) max. 0.5 %
Sodium (Na) (by AAS) max. 0.02 %
Sulfate (SO4) max. 0.001 %
Trace Impurities (in ppm)
Iron (Fe) max. 2
Nickel (Ni) max. 5
Heavy Metals (as Ag) max. 5
Nitrogen Compounds (as N) max. 5
Phosphate (PO4) max. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR