Test kits, photometric, Nitrite, LCK cuvette tests
Toimittaja: Hach
LCK341 LCK342
705-3054EA 121 EUR
705-3054 705-3055
Test kits, photometric, Nitrite, LCK cuvette tests
Ympäristöanalytiikan pikatestit
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C
Sertifikaatit: EN ISO 26777, DIN 38405 D10