Test kits, photometric, Cadmium, LCK cuvette tests
Toimittaja: Hach
LCK308
705-3016EA 160 EUR
705-3016
Test kits, photometric, Cadmium, LCK cuvette tests
Ympäristöanalytiikan pikatestit
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C (keep refrigerated)