Test kits, photometric, Alcohol, LCK cuvette tests
Toimittaja: Hach
LCK300
705-3002EA 212 EUR
705-3002
Test kits, photometric, Alcohol, LCK cuvette tests
Ympäristöanalytiikan pikatestit
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C (keep refrigerated)