Generic pocket colorimeters, DR300
Toimittaja: Hach
LPV4459960110 LPV4459950110 LPV4459952110 LPV4459965110
705-1179EA 959 EUR
705-1179 705-1177 705-1178 705-1180
Generic pocket colorimeters, DR300
Kolorimetrit Yksiparametri-kolorimetrit
Generic pocket colorimeter DR300 instruments are intended for customised applications not included in standard analysis systems. They are available in a range of wavelengths appropriate for many custom calibrations. In addition to the factory absorbance calibration (0 to 2,5 A), the instruments offer two user-entered, two-point linear calibrations, and provide a choice of either 0,00 to 2,55 or 0,0 to 99,0 for the display value.
  • Single parameter
  • Waterproof, lightweight, rugged
  • Backlit display
  • Data logging recall function
These instruments accept twoMore Product Information