Visible spectrophotometers, DR3900
Toimittaja: Hach
LPV4409900001 LPV4409800001 LPV4409910001
705-0764EA 7420 EUR
705-0764 705-1062 705-1147
Visible spectrophotometers, DR3900
Spektrofotometrit ja fotometrit UV/Vis-spektrofotometrit
Visible spectrophotometer to ensure the reliability and traceability of measurement results during routine analysis.
  • Integrated barcode reader (IBR+) increases the reliability of measurement values
  • Data updates and quality assurance with AQA+
  • Alignment of laboratory and process analysis by LINK2SC
  • Key methods are possible including COD, chlorine, nitrates and ammonia
  • High quality reagents formulated to ensure outstanding performance and results
  • Transmittance (%) - absorbance - concentration - scanning
Radio frequency identification (RFID) guaranteesMore Product Information