Pocket colorimeters, DR300
Toimittaja: Hach
LPV4459901110 LPV4459902110 LPV4459903110 LPV4459904110 LPV4459906110 LPV4459909110 LPV4459910110 LPV4459912110 LPV4459915110 LPV4459916110 LPV4459922110 LPV4459925110 LPV4459926110 LPV4459940110 LPV4459951110 LPV4459962110 LPV4459900110
705-1129EA 932 EUR
705-1129 705-1130 705-1131 705-1132 705-1133 705-1134 705-1135 705-1136 705-1137 705-1138 705-1139 705-1140 705-1162 705-1141 705-1142 705-1143 705-1128
Pocket colorimeters, DR300
Kolorimetrit Yksiparametri-kolorimetrit
The DR300 colorimeter is a reliable instrument that provides state-of-the-art data transfer capability and connection to Claros. Simple, intuitive operation reduces potential manual error, ensuring accurate measurement data.
  • Clear readings: Even under difficult conditions, thanks to backlit display and large readout
  • Waterproof, lightweight and rugged
The rugged, waterproof (IP 67) design withstands difficult conditions that can be encountered in the field (drops, extreme temperatures, rain and dirt), and still delivers dependable, accurateMore Product Information