Photometer, MD 610
Toimittaja: Lovibond
214025
705-0923EA 2350 EUR
705-0923
Photometer, MD 610
Kolorimetrit Moniparametri-Kolorimetrit
The MD 610 combines the mobility of a handheld photometer with the characteristics of a modern laboratory photometer. This instrument has over 120 pre-programmed methods covering the important methods for water analysis, including COD, chlorine, nutrients and metals. The high level of accuracy of Lovibond® reagents and the user friendly nature of the instrument guarantee rapid and reliable analysis of your water samples. Depending on the application, the unit will operate with tablet reagents, powder packs, liquid reagents or tube tests (¹⁶/₁₃ mm).
  • Bluetooth® data transmittance to