Test kits, chlorine, Comparator 2000+
Toimittaja: Lovibond
411120 411130 411250 411125 411126 411128
705-0351EA 1050 EUR
705-0351 705-0352 705-0356 LOVI411125 LOVI411126 LOVI411128
Test kits, chlorine, Comparator 2000+
Ympäristöanalytiikan pikatestit
Comparator 2000+ is a versatile modular system for testing water. Applicable in pool-water, research centres, universities, special applications, laboratory and field testing.
  • Compact and portable
  • Provide accurate and reproducible results
  • Colour-stable, fade-free glass standards
  • More than 400 different test discs are available
  • Compensation for coloured and turbid samples
  • Guaranteed constancy of the coloured glass standards
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
Sertifikaatit: In accordance with ISO 7393/2 'Determination of free chlorine and total chlorine'.
Toimitustiedot: Kit is supplied in a robust carrying case containing comparator 2000+, chlorine comparator disc, 4 x comparator vials, 10ml, 13mm path length, tablet reagents, crushing/stirring rod, vial cleaning brush and instruction sheet.