Test strips, chlorine, free and total
Toimittaja: Hach
2745050 2890200 2793944
705-0187EA 36.1 EUR
705-0187 HACH2890200 HACH2793944
Test strips, chlorine, free and total
Ympäristöanalytiikan pikatestit
Test Strips that allow technicians in the field to test many samples in only a few minutes, and make immediate evaluations on-site.
  • Easy to use
  • Disposable
  • Inexpensive
  • Quick, quantitative answers in the field or in the lab
The most widely used disinfectant for drinking water, chlorine is also important for sanitising swimming pools, cooling towers, other industrial equipment, and in the treatment of municipal wastewater. Its measurement and control are vital for both safety and economic reasons. Chlorine exists in water as 'free' chlorine (hypochlorite) or 'combined' chlorine (chloramines). The total chlorine test detects both forms. No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing.