Distillation, extraction and thermal reactions, behrotest® DET systems
Toimittaja: BEHR
B00218114
705-0023EA 8030 EUR
705-0023
Distillation, extraction and thermal reactions, behrotest® DET systems
Hajotusliuosjärjestelmät Mineralisointityöasemat
The behrotest® universal analytical DET system is a flexible modular system designed for laboratories with frequently changing applications who do not want to invest in dedicated complete apparatus for each application. The system has a core or basic unit to which application specific supplementary/expansion sets can be added. The expansion kits comprise typically of the additional specific glassware and gas flow connections.
The behrotest® DET DBAS includes the following equipment: