Groundwater sampling capsules, Polycap™ GW, Whatman™
Toimittaja: Whatman products (Cytiva)
6724-6004 6714-6004
701-0173EA 7440 EUR
701-0173 701-0164
Groundwater sampling capsules, Polycap™ GW, Whatman™
Suodattimet Membraanisuodatinkapselit
These Polycap™ GW (Ground Water) capsule filters are specifically designed for sample preparation of ground water samples for dissolved metals analysis. The in-line design with stepped hose barbs and hydrophilic polyethersulfone (PES) membrane make it easy to use for field sampling.
  • Stepped hose barb fittings allow for connection directly to outlet of a sampling pump with various size tubing
  • Effective filtration surface area of 600 cm² to ensure rapid sample collection
  • PES filters in thermally fused housing eliminate sample contamination that could come