VWR® Real Time Electrophoresis Systems
Toimittaja: VWR Collection
700-1472EA 939 EUR
700-1472 700-1473
VWR® Real Time Electrophoresis Systems
Elektroforeesi Vaakasuorat geelielektroforeesijärjestelmät
VWR® Real Time electrophoresis systems have revolutionised DNA electrophoresis. it is no longer necessary to remove a gel from a tank, place it onto a transilluminator for viewing or place the gel into an imaging system to take a photo. These versatile and compact systems are an all-in-one replacement for a power supply, gel tank, transilluminator and gel imaging system.
  • Monitor DNA separation in real time
  • Blue LED transilluminator (included) slides under the gel tank
  • Amber emission filter is built into the gel tank safety lid
  • Imaging enclosure included