VWR® PerfectBlue, Power Supply, High Current
Toimittaja: VWR Collection
700-1316EA 1100 EUR
700-1316
VWR® PerfectBlue, Power Supply, High Current
Virtalähteet
VWR® PerfectBlue power supply is constructed with flame-retardant ABC face plate and is ideal for DNA, RNA, and protein electrophoresis, as well as blotting and all routine horizontal and vertical electrophoresis.
  • Compact size, with a stackable case, provides a small footprint
  • TFT-LCD (2,4') colour screen shows all parameters during operation
  • Timer with alarm function
  • Constant voltage/current/power operation mode
  • Restart the operating program automatically after power outage
The power supply is microprocessor controlled and has four pairs of outletMore Product Information