Kaliumhydroksidi ≥85%, pellets, AR® ACS, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials
Vaara

Synonyms: Kalilipeä

6984-28EA 0 EUR
6984-28 6984-20
Kaliumhydroksidi ≥85%, pellets, AR® ACS, Macron Fine Chemicals™
Kaliumhydroksidi

Pellets.

Kaava: KOH
MW: 56.11 g/mol
Kiehumispiste: 1320
Sulamispiste: 360
Tiheys: 2.04 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003553
CAS-numero: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Testitulosten erittely

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 86 %
Assay (KOH) ≥85 %
Identification Passes Test
Calcium (Ca) ≤0.005 %
Chloride (Cl) ≤0.01 %
Heavy Metals (as Ag) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Nickel (Ni) ≤0.001 %
ACS - Magnesium (Mg) ≤0.002 %
Nitrogen Compounds (as N) ≤0.001 %
ACS - Phosphate (PO₄) ≤5 ppm
Sodium (Na) ≤0.05 %
Potassium Carbonate (K₂CO₃) ≤2.0 %
ACS - Sulfate (SO₄) ≤0.003 %
Appearance (white hygroscopic pellets)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR