Supelco® TraceCERT® Organic Reference Standard, Butyraldehyde-2,4-DNPH
Toimittaja: Merck
69630-50MG
69630-50MGEA 253 EUR
69630-50MG
Supelco® TraceCERT® Organic Reference Standard, Butyraldehyde-2,4-DNPH
Standardiaineet ja vertailumateriaalit Organic Standards
  • GHS signal: GHS07
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Varoitus: P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4807a39639629df5b44bc9276e056ee3ef60f215&vid=4807a39639629df5b44bc9276e056ee3ef60f215&prid=web&clid=SAPDAM