Muurahaishappo ≥96% ACS, Fluka™

Toimittaja: Honeywell Chemicals
Vaara

695076-100ML 695076-2.5L 695076-500ML
695076-100MLEA 32.6 EUR
695076-100ML 695076-2.5L 695076-500ML
Muurahaishappo ≥96% ACS, Fluka™
Muurahaishappo
Kaava: HCO₂H
Kiehumispiste: 100,4 °C (1.013 hPa)
Sulamispiste: 4 °C
Tiheys: 1,22 g/cm3 (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003297
CAS-numero: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Vapor Pressure 44.8 mmHg ( 20 °C)
Assay (alkalimetric) Min. 96 %
Identity (IR) complying
Non-volatile matter Max. 0.003 %
Ammonium (NH₄) Max. 0.005 %
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.003 %
Sulfite (SO₃) complying
Acetic acid (CH₃COOH) Max. 0.4 %
APHA Max. 15
Appearance complying
Solution 1 mL + 3 mL (H₂O) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR