Tetrahydrofuraani 99,5 GC_ASSAY_METHOD, AR®, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials
Vaara

Synonyms: Butylene oxide, THF, Diethylene oxide

6778-45EA 0 EUR
6778-45 6778-25
Tetrahydrofuraani 99,5 GC_ASSAY_METHOD, AR®, Macron Fine Chemicals™
Tetrahydrofuraani
Kaava: C₄H₈O
MW: 72.11 g/mol
Kiehumispiste: 66
Sulamispiste: -108.5
Leimahduspiste: –14 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005356
CAS-numero: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (GC) ≥99.5%
Water (KF ≤0.05 %
Titrable Acid (meq/g) ≤0.0005
Residue after Evaporation ≤0.0005 %
Peroxides (as H₂O₂) ≤0.015 %
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR