Rare earth element mix for ICP, 16 elements, TraceCERT®, Supelco®
Toimittaja: Merck
67349-100ML
67349-100MLEA 333 EUR
67349-100ML
Rare earth element mix for ICP, 16 elements, TraceCERT®, Supelco®
Standardiaineet ja vertailumateriaalit Cationic Standards Multi-Element Standards
50 mg/l each: Sc, Y, La, Ce, Pr, Nd, Sm, Eu, Gd, Tb, Dy, Ho, Er, Tm, Yb and Lu in 2% nitric acid
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034