Multi-parameter meter (pH/ISE/conductivity/DO), bench, inoLab® Multi 9630 IDS
Toimittaja: WTW
1FD570 1FD57K
665-0487EA 3890 EUR
665-0487 665-0525
Multi-parameter meter (pH/ISE/conductivity/DO), bench, inoLab® Multi 9630 IDS
Sähkökemiamittarit pH-mittarit
Digital high performance multi-parameter device with three galvanically isolated measuring channels for pH/mV, ISE (including incremental methods), conductivity and oxygen measurements. With comprehensive GLP functions.
  • Simultaneous measurement with three independent measuring channels
  • User-defined combinations of equal or different parameters
  • Continuous Measurement Control (CMC) for measuring in the optimum pH range
  • Quality Sensor Control (QSC) benchmarks the current condition of the special IDS electrodes
  • Digital processing and data transfer (GLP