Multi-parameter meter (pH/mV/conductivity), bench, Orion Star™ A215
Toimittaja: Thermo Orion
STARA2155 STARA2150
665-0272EA 2320 EUR
665-0272 665-0271
Multi-parameter meter (pH/mV/conductivity), bench, Orion Star™ A215
Sähkökemiamittarit pH-mittarit
This benchtop meter is ideal for a wide range of applications and advanced pH and conductivity analysis in the lab. It offers two measuring channels allowing users to measure pH and conductivity simultaneously or view each channel separately.
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/