Oxygen Sensor, Memosens COS81E
Toimittaja: Endress+Hauser
COS81E-10L1/0 COS81E-14W2/0 COS81E-14Q0/0 COS81E-14V7/0
664-0243EA 3290 EUR
664-0243 664-0244 664-0245 664-0246
Oxygen Sensor, Memosens COS81E
Elektrodit ja niiden oheistarvikkeet Liuennut happi
Memosens 2.0 hygienic sensor for the life sciences and food industries.
  • Memosens COS81E is designed according to EHEDG and ASME BPE. It avoids cross-contamination and is fully compliant to USP class VI and FDA as well as all GMP and GLP requirements
  • High process up-time: Precalibrate the sensor in your lab and then swap it into your process with plug and play. It does not need polarization time and is immediately ready to measure
  • Reliable measurement: A built-in reference LED compensates the ageing of the measuring LED. This ensures precise measured values