Johtokykymittari, bench, FiveEasy™ F30 / FiveEasy™ Plus FP30
Toimittaja: Mettler - Toledo
30266942 30266943 30266944 30266945
663-0295EA 1030 EUR
663-0295 663-0296 663-0297 663-0298
Johtokykymittari, bench, FiveEasy™ F30 / FiveEasy™ Plus FP30
Sähkökemiamittarit Johtokykymittarit
These bench top instruments provide high quality conductivity measurements with the simple click of a button. Thanks to their intuitive operation, sleek design and simple data export functions they are easy to set up and use. Ideal for use in food and beverage processing, agriculture, environment, industry and for education purposes.
  • Robust ABS housing and large, well-structured display for easy viewing
  • Compact design: Requires very little bench space in the lab
  • Sensor holder can be easily removed and stored on the side of the meter after use
  • Acoustic and