Johtokyky, mittauskennot, InLab® ISM®
Toimittaja: Mettler - Toledo
51344110 51344112 51344114 51344116 51344118 30014094 30014092 30014093
663-0000EA 943 EUR
663-0000 663-0268 663-0269 663-0145 663-0270 663-0178 663-0176 663-0177
Johtokyky, mittauskennot, InLab® ISM®
Elektrodit ja niiden oheistarvikkeet Johtokyky
High performance sensors combining robustness with state-of-the-art measurement technology. Compatible with either SevenExcellence™ and SevenCompact™ meters (663-0176/663-0177/663-0178) or with SevenGo Duo™ SG23 and SevenGo Duo pro™ SG78 meters (663-000/663-0145). Fully traceable.
The revolutionary intelligent sensor management system (ISM®) offers the user an even greater level of security. The calibration data is stored on a chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.