Elektrodi, pH-elektrodit, InLab®, Expert-sarja
Toimittaja: Mettler - Toledo
51340288 51343100 51343101 51343102 51343103 51343104 51343105
662-2738EA 547 EUR
662-2738 662-2839 662-2840 662-2841 662-2842 662-2843 662-2844
Elektrodi, pH-elektrodit, InLab®, Expert-sarja
Elektrodit ja niiden oheistarvikkeet pH-elektrodit
Ideal for critical samples such as suspensions or samples of unknown composition (e.g. wastewater).
  • ARGENTHAL™ reference system with silver-ion trap
  • Solid XEROLYT® polymer electrolyte: No need for a junction (no possibility of contamination)
  • Unbreakable PEEK shaft
  • Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
pH-alue: 0 to 14
Temperature range: 0 to 100 °C
Lämpötila-anturi: NTC30 kOhm malleissa InLab® Expert Pro ja InLab® Expert NTC30, PT1More Product Information