Elektrodi, pH-yhdistelmäelektrodit
Toimittaja: Mettler - Toledo
00089632 51109502 51109504 51109502
662-2736EA 491 EUR
662-2736 662-2160 662-1669 662-2478
Elektrodi, pH-yhdistelmäelektrodit
Elektrodit ja niiden oheistarvikkeet pH-elektrodit
Combined glass pH electrodes for direct pH measurement and acid/base titrations in aqueous or non-aqueous media. The all-rounder for a large variety of uncritical applications in almost all kinds of industries: Acid content of beverages, acid and base capacity of water samples, pH start applications.
  • Integrated glass sleeve for secure fixation in the titration head
  • With S7 screw cap and detachable cap or with Plug & Play S12 connection
  • Plug & Play sensor features an integrated sensor chip which allows data storing and monitoring
DG111-SC/DGi111More Product Information