VWR® pH 2500L, pH/mV/°C Bench Meter, with Colour Graphic Touch Screen
Toimittaja: VWR Collection
662-2380EA 972 EUR
662-2380
VWR® pH 2500L, pH/mV/°C Bench Meter, with Colour Graphic Touch Screen
Sähkökemiamittarit pH-mittarit
The user friendly instrument is distinguished by a large a 127 mm colour graphic touch screen. The meter can be used for accurate measurements of pH, mV and temperature. This pH meter enables simultaneous measurements displaying and storing of two functions (pH and temperature).
  • Large five colour graphic touch screen showing both pH/mV and temperature value simultaneously
  • 1- to 3-point calibration with automatic detection of buffer solutions and NIST standard solutions
  • Storing characteristics of three electrodes enables their quick replacement
  • Data-logger