Kalorimetri, C 6000 global standards and isoperibol
Toimittaja: IKA
8804300 8804400 8804700 8804800 10004520 10004521 10004522 10004523 10004524 10004525 10004526 10004527
IKAA8804300EA 45760 EUR
IKAA8804300 IKAA8804400 IKAA8804700 IKAA8804800 662-2321 662-2322 662-2323 662-2324 662-2325 662-2326 662-2327 662-2328
Kalorimetri, C 6000 global standards and isoperibol
Kalorimetrit
The C 6000 global standards and C 6000 isoperibol calorimeters follow the approved C 5000 and C 2000 calorimeter models.
  • Convenient touch screen operation
  • Software provides control chart view and correction calculation
  • RFID technology used for decomposition vessel identification
  • SD card slot, Ethernet interface for additional data management
  • Decomposition vessel with spherical top for faster heat transfer results in shorter experimental times
  • Easy bomb preparation due to 'turned around' crucible holder technology
The C 6000 global standards offers aMore Product Information