VWR® pHenomenal® IDP pH Electrodes
Toimittaja: VWR Collection
662-2297EA 357 EUR
662-2297 662-2296 662-2298
VWR® pHenomenal® IDP pH Electrodes
Elektrodit ja niiden oheistarvikkeet pH-elektrodit
These IDP (Intelligent Digital Probe) pH/temperature electrodes can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.
  • Calibration data (serial number, calibration status) is stored in the electrode/probe, transmitted to the meter and displayed
  • Proven reliability and accuracy
  • Correct measurement results due to direct signal processing in the sensor and digital transmission of values
  • All parameters are easily measured with cable lengths of up to 100 metres
  • Same universal connector for all electrodes and probes
TemperatureMore Product Information