pH-mittari, moniparametrimittarit, kädessä pidettävä, MultiLine® Multi 3510 IDS
Toimittaja: WTW
2FD350 2FD351 2FD353 2FD354 2FD35U
662-2135EA 1240 EUR
662-2135 662-2136 662-2137 662-2138 662-2139
pH-mittari, moniparametrimittarit, kädessä pidettävä, MultiLine® Multi 3510 IDS
Sähkökemiamittarit pH-mittarit
This compact portable multi-parameter instrument enables measurements of pH, conductivity and dissolved oxygen with intelligent digital sensors (IDS). It has a universal measurement channel for different parameters. The robust, waterproof design makes it ideal for measurements in the field and in the laboratory.
  • Well laid out menus make operation safe and easy
  • MultiLab® Importer software enables data acquisition via Excel®
  • QSC function for optimum tracking of sensor quality
  • Calibration records and additional information are stored in the sensor
  • Wireless-