Elektrodi, ORP-elektrodi, SenTix® ORP-T 900, IDS
Toimittaja: WTW
103791
662-2020EA 408 EUR
662-2020
Elektrodi, ORP-elektrodi, SenTix® ORP-T 900, IDS
Elektrodit ja niiden oheistarvikkeet Redox Electrodes
IDS ORP electrodes are used to measure oxidation or reduction potentials in aqueous solutions - with integrated temperature sensor.
  • Safe measured values due to signal processing in the sensor
  • Integrated temperature sensor NTC 30 kOhm for precise value documentation
  • Best possible GLP support by documenting the sensor data
Platinum IDS ORP electrode with liquid electrolyte and glass shaft, temperature sensor, ceramic junction.