Elektrodi, pH-yhdistelmäelektrodit, IntelliCAL™
Toimittaja: Hach
PHC10101 PHC10103 PHC10105 PHC10110 PHC10115 PHC10130 PHC30101 PHC30103 PHC28103 PHC28101 PHC20101 PHC20103
662-1982EA 304 EUR
662-1982 662-1983 662-1984 665-0418 HACHPHC10115 662-1985 662-1989 HACHPHC30103 662-1988 662-1987 662-1986 HACHPHC20103
Elektrodi, pH-yhdistelmäelektrodit, IntelliCAL™
Elektrodit ja niiden oheistarvikkeet pH-elektrodit
These digital combination electrodes feature a built-in temperature sensor and are either refillable or non refillable. They can be used with all Hach HQd™ meters.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Ideal for a multi-user environment with multiple HQd series meters
  • Ultimate traceability: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • Alert the user when re-calibration is needed
  • Electrodes can be moved between meters without the need to re-calibrate or