VWR® pH 110/pH 110 M, pH-mittari, pH/mV/°C-mittari, kädessä pidettävä
Toimittaja: VWR Collection
662-1350EA 611 EUR
662-1350 662-2268 662-2269 662-2270
VWR® pH 110/pH 110 M, pH-mittari, pH/mV/°C-mittari, kädessä pidettävä
Sähkökemiamittarit pH-mittarit
These economical, robust and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carry case, electrode and temperature sensor with different cable lengths and buffers.
  • IP 67 waterproof housing, large display and easy to use keypad
  • Automatic/manual temperature compensation
  • 50 or 250 data sets reviewable memory
  • 1000 hour battery life; auto-shut off function (30 minutes of non use)
  • Electrode offset recognition; auto-lock and hold of readings on display
Additional functions of pH 110 M:
- Extended memory capacityMore Product Information