VWR® pHenomenal®, Elektrodi, pH-elektrodi
Toimittaja: VWR Collection
620-1659EA 186 EUR
620-1659 662-1162 662-1163 662-1164 662-1158 662-1159 662-1161 662-1156 662-1157 662-1247
VWR® pHenomenal®, Elektrodi, pH-elektrodi
Elektrodit ja niiden oheistarvikkeet pH-elektrodit
pH-elektrodit BNC-liittimellä ja kaapelilla.
These electrodes are checked at five points during the manufacturing process and then lot stamped to ensure quality. Every electrode is supplied with a soaker storage bottle and cap to ensure the pH-sensitive glass and reference junction remain in top condition and are instantly ready for use.
* Integrated temperature probe.
Toimitustiedot: Includes pH-electrode only. 620-1659 includes pHenomenal® TEMP21 T-sensor (NTC 30 kOhm) only.