Dissolved oxygen probes, Orion™
Toimittaja: Thermo Orion
083010MD 083005MD
662-1136EA 1030 EUR
662-1136 664-0152
Dissolved oxygen probes, Orion™
Elektrodit ja niiden oheistarvikkeet Liuennut happi
These standard DO probes can be used in the lab or in the field.
  • Durable and accurate
  • Sensor types: Dual thermally separated
  • Automatic temperature compensation
  • Convenient screw-on membrane caps
  • Response time: 90% of final value in 10 sec.
DO probe Orion 083005A is for use with Orion 830A, 835A meters. Cable length: 1,5 m, 8-pin waterproof DIN connection.
DO Probe Orion 083010A is for use with Orion 862A meter. Cable length: 3 m, 8-pin waterproof DIN connection.
DO Probe Orion 083005D is for use with Orion 810Aplus, 850Aplus meters. Cable length: 1,5 m, 8More Product Information