Orion™ Gas-Sensing ISE Membranes
Toimittaja: Thermo Orion
951204 950204 951205 951215
662-0856EA 170 EUR
662-0856 ORIO950204 662-1515 662-1671
Orion™ Gas-Sensing ISE Membranes
Elektrodit ja niiden oheistarvikkeet Ioniselektiiviset elektrodit, ISE
These fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
  • Loose or preassembled membranes
  • Easy to use