pH electrode storage and cleaning solutions, Orion™

Toimittaja: Thermo Orion

900022 900020 910060 9100CB 911110 900023 900024 910001 911125
662-0757EA 148 EUR
662-0757 662-0979 662-1552 ORIO9100CB ORIO911110 662-0758 662-0759 662-0768 662-0783
pH electrode storage and cleaning solutions, Orion™
Elektrolyytit Elektrodin säilytysliuokset

Solutions for storage and cleaning that prolong the working life of pH electrodes. Cleaning solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation.


  • Cleaning solution A: for removing protein contaminants
  • Cleaning solution B: for removing bacterial contaminants
  • Cleaning solution C: for general cleaning
  • Cleaning solution D: for removing oil and grease contaminants


Bottles include a small beaker to hold the solution and a plastic pipette to remove the old electrode filling solution.


Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR