Probe maintenance kits for dissolved oxygen meters, Orion™
Toimittaja: Thermo Orion
O80113 080513
662-0692EA 123 EUR
662-0692 664-0499
Probe maintenance kits for dissolved oxygen meters, Orion™
Elektrodit ja niiden oheistarvikkeet Liuennut happi
These probe maintenance kits help ensure optimal performance of Orion™ polarographic dissolved oxygen probes.
Toimitustiedot: Each probe maintenance kit includes two membrane caps, one electrolyte solution, and a polishing disk.