ISE sample conditioning buffers and solutions
Toimittaja: WTW
140140 106580 140100 140110 140120 150120 150125
662-0386EA 413 EUR
662-0386 662-0495 662-0359 662-0350 WTWA140120 662-0375 662-0379
ISE sample conditioning buffers and solutions
Biokemialliset puskuriliuokset Ionic Strength Adjustment Buffers
Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.